Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ube2q1
Entrez Gene ID
295252
Gene Name
ubiquitin-conjugating enzyme E2Q family member 1
Gene Aliases
Ube2q
Chromosome Location
Chr.2: 189106039 - 189115110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106448.2 NP_001099918.1 8 - 9 1052 79
GenBank mRNA BC166534.1 - 8 - 9 1052 79

View Full Product Details