Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slx4
Entrez Gene ID
302953
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
Btbd12, RGD1566009
Chromosome Location
Chr.10: 11787756 - 11808331 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245835.2 XP_006245897.2 5 - 6 1199 76

View Full Product Details