Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ksr2
Entrez Gene ID
288691
Gene Name
kinase suppressor of ras 2
Gene Aliases
RGD1562674
Chromosome Location
Chr.12: 44531376 - 44911824 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249485.2 XP_006249547.1 6 - 7 1919 64

View Full Product Details