Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsd3b
Entrez Gene ID
682974
Gene Name
3 beta-hydroxysteroid dehydrogenase/delta-5-delta-4 isomerase type II
Gene Aliases
-
Chromosome Location
Chr.2: 200756557 - 200762492 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042619.1 NP_001036084.1 2 - 3 295 157
GenBank mRNA M38179.1 - 2 - 3 295 157

View Full Product Details