Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oxgr1
Entrez Gene ID
290493
Gene Name
oxoglutarate (alpha-ketoglutarate) receptor 1
Gene Aliases
Gpr80, P2Y15
Chromosome Location
Chr.15: 105399596 - 105422534 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207588.1 NP_997471.1 1 - 1 964 155
XM_006252471.2 XP_006252533.1 2 - 2 1214 155
GenBank mRNA AY191367.1 - 1 - 1 964 155

View Full Product Details