The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106223.1 | NP_001099693.1 | 3 - 3 | 2438 | 141 |
XM_006228252.2 | XP_006228314.1 | 3 - 3 | 2546 | 141 | |
XM_006228253.2 | XP_006228315.1 | 3 - 3 | 2393 | 141 | |
XM_006228254.2 | XP_006228316.1 | 3 - 3 | 2978 | 141 | |
GenBank mRNA | FQ224110.1 | - | 3 - 3 | 2378 | 141 |
FQ224124.1 | - | 3 - 3 | 2371 | 141 |