The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113751.1 | NP_001107223.1 | 1 - 2 | 282 | 70 |
GenBank mRNA | BC158683.1 | - | 1 - 2 | 282 | 70 |