Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PET9
Entrez Gene ID
852250
Gene Name
Gene Aliases
AAC2, ANC2, YBL030C
Chromosome Location
Chr.II: 163041 - 163997 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178270.1 NP_009523.1 1 - 1 850 69

View Full Product Details