Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UMP1
Entrez Gene ID
852471
Gene Name
Gene Aliases
RNS2, YBR173C
Chromosome Location
Chr.II: 581726 - 582172 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178521.1 NP_009732.1 1 - 1 346 92

View Full Product Details