Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP31
Entrez Gene ID
852146
Gene Name
Gene Aliases
YDR533C
Chromosome Location
Chr.IV: 1501447 - 1502160 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180841.1 NP_010822.1 1 - 1 649 67

View Full Product Details