Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP25
Entrez Gene ID
855123
Gene Name
Gene Aliases
YMR098C
Chromosome Location
Chr.XIII: 460771 - 462609 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182598.1 NP_013816.1 1 - 1 1655 104

View Full Product Details