Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCF2
Entrez Gene ID
855752
Gene Name
Gene Aliases
AIM38, YNR018W
Chromosome Location
Chr.XIV: 664270 - 664944 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183195.1 NP_014415.1 1 - 1 552 88

View Full Product Details