Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPT1
Entrez Gene ID
854384
Gene Name
Gene Aliases
YOR209C
Chromosome Location
Chr.XV: 736437 - 737726 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183628.1 NP_014852.1 1 - 1 1047 92

View Full Product Details