Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMC5
Entrez Gene ID
397043
Gene Name
proteasome 26S subunit, ATPase 5
Gene Aliases
TBP10
Chromosome Location
Chr.12: 15101946 - 15106326 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213983.1 NP_999148.1 10 - 11 1230 145
XM_005668681.3 XP_005668738.1 - 1313 145
XM_013980839.2 XP_013836293.1 - 1362 145
GenBank mRNA AK234194.1 - 10 - 11 1120 145
AK395258.1 - 10 - 11 1142 145
AK396074.1 - 10 - 11 1115 145
X89719.1 - - 1058 145

View Full Product Details