Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGP
Entrez Gene ID
397206
Gene Name
matrix Gla protein
Gene Aliases
MPG
Chromosome Location
Chr.5: 57665272 - 57669360 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214116.1 NP_999281.1 2 - 3 140 100
GenBank mRNA AF525316.1 - 2 - 3 140 100
AK346558.1 - 2 - 3 159 100
GU391766.1 - 2 - 3 159 100

View Full Product Details