Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC5A4
Entrez Gene ID
397376
Gene Name
solute carrier family 5 member 4
Gene Aliases
PSGLT2
Chromosome Location
Chr.14: 48708047 - 48741440 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214182.1 NP_999347.1 14 - 15 1769 70
GenBank mRNA L02900.1 - 14 - 15 1769 70

View Full Product Details