Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPPA
Entrez Gene ID
397496
Gene Name
natriuretic peptide A
Gene Aliases
ANF, ANP, CDD, CDP
Chromosome Location
Chr.6: 71919557 - 71921345 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214260.2 NP_999425.1 1 - 2 132 70
XM_005664986.3 XP_005665043.1 - 261 70
GenBank mRNA X54669.1 - - 133 70

View Full Product Details