Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA3
Entrez Gene ID
100415932
Gene Name
sperm acrosome associated 3
Gene Aliases
-
Chromosome Location
Chr.12: 42046804 - 42050684 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177928.1 NP_001171399.1 3 - 4 423 92
GenBank mRNA GU474991.1 - 3 - 4 423 92

View Full Product Details