Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CATIP
Entrez Gene ID
100155612
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C15H2orf62, C2orf62
Chromosome Location
Chr.15: 120419473 - 120425441 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190220.1 NP_001177149.1 8 - 9 875 91
GenBank mRNA AK234312.1 - 8 - 9 850 91
AK238360.1 - 8 - 9 875 91

View Full Product Details