Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A7
Entrez Gene ID
100628141
Gene Name
membrane spanning 4-domains A7
Gene Aliases
-
Chromosome Location
Chr.2: 11295925 - 11320284 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243358.1 NP_001230287.1 6 - 7 954 92
XM_021080021.1 XP_020935680.1 - 1089 92
GenBank mRNA AK231277.1 - 6 - 7 934 92
AK240438.1 - 6 - 7 937 92
CB479849.1 - 1 - 2 175 92

View Full Product Details