Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P5CS
Entrez Gene ID
100232881
Gene Name
pyrroline-5-carboxylate synthetase
Gene Aliases
-
Chromosome Location
Chr.13: 3718569 - 3736708 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ005686.1 - 3 - 4 285 86

View Full Product Details