Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hid1
Entrez Gene ID
395008
Gene Name
HID1 domain containing
Gene Aliases
c17orf28
Chromosome Location
Chr.10: 35676362 - 35694555 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204046.1 NP_989377.1 5 - 6 721 67
XM_012971610.2 XP_012827064.1 5 - 6 776 67
GenBank mRNA BC064723.1 - 5 - 6 721 67

View Full Product Details