Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab3d
Entrez Gene ID
448064
Gene Name
RAB3D, member RAS oncogene family
Gene Aliases
d2-2, gov, rab16, rab3a, rab3d-a, rab3d-b, rad3d
Chromosome Location
Chr.3: 129090755 - 129105110 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005621.1 NP_001005621.1 3 - 4 464 70
GenBank mRNA BC074589.1 - 3 - 4 464 70
CT025365.2 - 3 - 4 420 70

View Full Product Details