Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spa17
Entrez Gene ID
448437
Gene Name
sperm autoantigenic protein 17
Gene Aliases
-
Chromosome Location
Chr.7: 64228858 - 64303739 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006757.1 NP_001006758.1 3 - 4 319 83
XM_012966399.2 XP_012821853.1 3 - 4 590 83
XM_012966400.2 XP_012821854.1 3 - 4 705 83
GenBank mRNA BC075539.1 - 3 - 4 319 83
CT030312.1 - - 369 83

View Full Product Details