Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eci1
Entrez Gene ID
496654
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
dci
Chromosome Location
Chr.9: 14352528 - 14361113 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011219.1 NP_001011219.1 6 - 7 738 77
GenBank mRNA BC087779.1 - 6 - 7 738 77

View Full Product Details