Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rp9
Entrez Gene ID
496684
Gene Name
retinitis pigmentosa 9 (autosomal dominant)
Gene Aliases
-
Chromosome Location
Chr.6: 26102786 - 26115882 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011242.1 NP_001011242.1 6 - 7 578 116
XM_012964601.2 XP_012820055.1 6 - 7 744 116
GenBank mRNA BC087814.1 - 6 - 7 578 116
CR761073.2 - 6 - 7 565 116

View Full Product Details