Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyb5d2
Entrez Gene ID
548908
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 97511333 - 97517336 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016154.1 NP_001016154.1 1 - 2 300 110
GenBank mRNA BC161279.1 - 1 - 2 284 110
BC170615.1 - 1 - 2 296 110
BC170621.1 - 1 - 2 296 110
CR761951.1 - 1 - 2 300 110

View Full Product Details