Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyb5d2
Entrez Gene ID
548908
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 97511333 - 97517336 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016154.1 NP_001016154.1 2 - 3 445 102
GenBank mRNA BC161279.1 - 2 - 3 429 102
BC170615.1 - 2 - 3 441 102
BC170621.1 - 2 - 3 441 102
CR761951.1 - 2 - 3 445 102

View Full Product Details