Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
armc6
Entrez Gene ID
448573
Gene Name
armadillo repeat containing 6
Gene Aliases
-
Chromosome Location
Chr.1: 102121202 - 102130425 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005042.2 NP_001005042.1 4 - 5 957 79
XM_012960033.2 XP_012815487.1 5 - 6 1142 79
GenBank mRNA BC076906.1 - 5 - 6 1013 79
CR926276.2 - 4 - 5 957 79

View Full Product Details