Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
reep6
Entrez Gene ID
548805
Gene Name
receptor accessory protein 6
Gene Aliases
dp1l1
Chromosome Location
Chr.1: 80450051 - 80456856 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016051.2 NP_001016051.1 5 - 6 624 60
GenBank mRNA BC122013.1 - 5 - 6 625 60
CR762344.2 - 5 - 6 624 60

View Full Product Details