Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrpl40
Entrez Gene ID
549003
Gene Name
mitochondrial ribosomal protein L40
Gene Aliases
-
Chromosome Location
Chr.1: 145725936 - 145731193 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016249.2 NP_001016249.1 3 - 4 411 98
GenBank mRNA BC135459.1 - 3 - 4 375 98
CR761757.2 - 3 - 4 411 98

View Full Product Details