Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ca12
Entrez Gene ID
549185
Gene Name
carbonic anhydrase XII
Gene Aliases
-
Chromosome Location
Chr.3: 81818749 - 81834713 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016431.2 NP_001016431.1 8 - 9 971 66
GenBank mRNA BC135174.2 - 8 - 9 904 66
CR761051.2 - 8 - 9 971 66

View Full Product Details