Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cab39l
Entrez Gene ID
549934
Gene Name
calcium binding protein 39 like
Gene Aliases
-
Chromosome Location
Chr.2: 146269746 - 146312017 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017180.2 NP_001017180.1 2 - 3 278 101
XM_012957037.1 XP_012812491.1 3 - 4 425 101
XM_012957038.2 XP_012812492.1 4 - 5 667 101
XM_012957040.2 XP_012812494.1 3 - 4 678 101
GenBank mRNA BC127354.1 - 3 - 4 358 101
CR760674.2 - 2 - 3 278 101

View Full Product Details