Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
msgn1
Entrez Gene ID
733924
Gene Name
mesogenin 1
Gene Aliases
Mespo, mes1, mesogenin, msgn1-a, msgn1-b, msog, pMesogenin1
Chromosome Location
Chr.5: 137752740 - 137753675 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045639.1 NP_001039104.1 1 - 1 451 58
GenBank mRNA BC075572.1 - 1 - 1 495 58
BC167882.1 - 1 - 1 499 58
CR942397.2 - 1 - 1 451 58

View Full Product Details