Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dhrs7b
Entrez Gene ID
779695
Gene Name
dehydrogenase/reductase (SDR family) member 7B
Gene Aliases
-
Chromosome Location
Chr.9: 33059889 - 33161093 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078778.1 NP_001072246.1 3 - 4 338 99
XM_012970534.2 XP_012825988.1 2 - 3 652 99
XM_012970535.2 XP_012825989.1 3 - 4 348 99
XM_012970536.2 XP_012825990.1 3 - 4 326 99
GenBank mRNA BC118856.1 - 3 - 4 338 99

View Full Product Details