Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp26b1
Entrez Gene ID
780112
Gene Name
cytochrome P450 family 26 subfamily B member 1
Gene Aliases
cyp26a2, p450rai-2
Chromosome Location
Chr.1: 8565371 - 8587702 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079187.2 NP_001072655.1 - 783 71
GenBank mRNA BC123940.1 - - 815 71
BC135551.1 - - 783 71

View Full Product Details