Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zc3h6
Entrez Gene ID
100124753
Gene Name
zinc finger CCCH type containing 6
Gene Aliases
zc3hdc6
Chromosome Location
Chr.5: 38972995 - 38988894 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102732.2 NP_001096202.2 4 - 5 804 92
GenBank mRNA BC135562.1 - 4 - 5 796 92

View Full Product Details