Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kansl2
Entrez Gene ID
100125116
Gene Name
KAT8 regulatory NSL complex subunit 2
Gene Aliases
c12orf41
Chromosome Location
Chr.2: 132378405 - 132393692 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103023.1 NP_001096493.1 2 - 3 443 97
XM_012956313.2 XP_012811767.1 3 - 4 616 97
XM_012956314.2 XP_012811768.1 3 - 4 517 97
XM_012956315.2 XP_012811769.1 3 - 4 818 97
XM_018091048.1 XP_017946537.1 3 - 4 746 97
XM_018091049.1 XP_017946538.1 3 - 4 487 97
GenBank mRNA BC135559.1 - 2 - 3 443 97

View Full Product Details