Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kansl2
Entrez Gene ID
100125116
Gene Name
KAT8 regulatory NSL complex subunit 2
Gene Aliases
c12orf41
Chromosome Location
Chr.2: 132378405 - 132393692 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103023.1 NP_001096493.1 7 - 8 1239 65
XM_012956313.2 XP_012811767.1 8 - 9 1412 65
XM_012956314.2 XP_012811768.1 8 - 9 1313 65
XM_012956315.2 XP_012811769.1 8 - 9 1614 65
XM_018091048.1 XP_017946537.1 8 - 9 1542 65
XM_018091049.1 XP_017946538.1 8 - 9 1283 65
GenBank mRNA BC135559.1 - 7 - 8 1239 65

View Full Product Details