Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crym
Entrez Gene ID
100125176
Gene Name
crystallin mu
Gene Aliases
-
Chromosome Location
Chr.9: 18890501 - 18917508 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103062.1 NP_001096532.1 5 - 6 715 87
XM_012969992.2 XP_012825446.1 5 - 6 672 87
XM_012969993.2 XP_012825447.1 6 - 7 763 87
XM_018097025.1 XP_017952514.1 6 - 7 637 87
XM_018097026.1 XP_017952515.1 5 - 6 657 87
XM_018097027.1 XP_017952516.1 5 - 6 549 87
GenBank mRNA BC135858.1 - 5 - 6 715 87

View Full Product Details