Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hyi
Entrez Gene ID
100135187
Gene Name
hydroxypyruvate isomerase (putative)
Gene Aliases
ht036
Chromosome Location
Chr.4: 74933123 - 74943016 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113890.1 NP_001107362.1 7 - 8 890 66
XM_012960563.2 XP_012816017.1 8 - 9 1436 66
XM_012960564.2 XP_012816018.1 8 - 9 1484 66
XM_012960565.1 XP_012816019.1 8 - 9 957 66
GenBank mRNA BC082746.1 - 8 - 9 959 66
BC157213.1 - 7 - 8 890 66
BC171074.1 - 7 - 8 783 66
BC171076.1 - 7 - 8 783 66

View Full Product Details