Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp1d1
Entrez Gene ID
100145341
Gene Name
cytochrome P450 family 1 subfamily D polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.1: 118488824 - 118502138 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126813.1 NP_001120285.2 3 - 4 1022 79
GenBank mRNA BC160554.1 - 3 - 4 1022 79

View Full Product Details