Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dusp11
Entrez Gene ID
100151717
Gene Name
dual specificity phosphatase 11
Gene Aliases
pir1
Chromosome Location
Chr.3: 113045692 - 113062278 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127420.1 NP_001120892.1 6 - 7 512 63
XM_012958615.2 XP_012814069.1 6 - 7 679 63
GenBank mRNA BC166270.1 - 6 - 7 512 63

View Full Product Details