Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rspo1
Entrez Gene ID
100158609
Gene Name
R-spondin 1
Gene Aliases
-
Chromosome Location
Chr.2: 59027556 - 59109701 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128028.1 NP_001121500.1 4 - 5 986 85
GenBank mRNA BC166324.1 - 4 - 5 986 85

View Full Product Details