Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs20
Entrez Gene ID
100158614
Gene Name
regulator of G-protein signaling 20
Gene Aliases
rgsz1, zgap1
Chromosome Location
Chr.6: 100004943 - 100038480 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128030.1 NP_001121502.1 4 - 5 370 101
GenBank mRNA BC166331.1 - 4 - 5 370 101

View Full Product Details