The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134804.1 | NP_001128276.1 | 2 - 3 | 234 | 65 |
XM_012962945.2 | XP_012818399.1 | 2 - 3 | 221 | 65 | |
GenBank mRNA | BC089077.1 | - | 2 - 3 | 201 | 65 |
BC168797.1 | - | 2 - 3 | 234 | 65 |