Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ribc1
Entrez Gene ID
100216275
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
-
Chromosome Location
Chr.8: 2270344 - 2307411 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142228.2 NP_001135700.1 2 - 3 206 100
XM_012968121.2 XP_012823575.1 3 - 4 302 100
XM_012968122.2 XP_012823576.1 3 - 4 303 100
XM_012968123.1 XP_012823577.1 3 - 4 304 100
XM_012968124.2 XP_012823578.1 3 - 4 225 100
GenBank mRNA BC168813.1 - 2 - 3 208 100
CR761888.2 - 3 - 4 326 100

View Full Product Details