Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MAGE
Entrez Gene ID
100194084
Gene Name
hypothetical protein
Gene Aliases
ZEAMMB73_271920
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139139.1 NP_001132611.1 - 1050 67
GenBank mRNA BT036524.1 - - 1050 67
BT038139.1 - - 838 67

View Full Product Details