Gene Transcripts
- Gene Symbol
- Sprr2g
- Entrez Gene ID
- 20761
- Gene Name
- small proline-rich protein 2G
- Gene Aliases
- Chromosome Location
- Chr. 3: 92373915 - 92375229 on Build GRCm38
- UniGene ID
- Mm.57083
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003548.1 |
|
2 |
617 |
|
GenBank mRNA |
AY158991.1 |
- |
2 |
616 |
|
View Full Product Details