Gene Transcripts

Gene Symbol
Afg3l1
Entrez Gene ID
114896
Gene Name
AFG3-like AAA ATPase 1
Gene Aliases
1700047G05Rik, 3110061K15Rik
Chromosome Location
Chr. 8: 123477862 - 123503916 on Build GRCm38
UniGene ID
Mm.287475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028260.1 13, 14 1685
GenBank mRNA AK168244.1 - 13, 14 1659
BC043320.1 - 4, 5 507
BC060064.1 - 13, 14 1644

View Full Product Details